Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190851.2 | NP_001177780.2 | 3 - 4 | 468 | 106 | |
XM_028830592.1 | XP_028686425.1 | 1856 | 106 |
Set Membership: |
![]() ![]() ![]() ![]() |