Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194481.2 | NP_001181410.1 | 5 - 6 | 1381 | 70 | |
XM_001110422.3 | XP_001110422.1 | 1381 | 70 | |||
XM_028832680.1 | XP_028688513.1 | 1317 | 70 | |||
XM_028832681.1 | XP_028688514.1 | 1403 | 70 | |||
XM_028832682.1 | XP_028688515.1 | 1317 | 70 | |||
GenBank mRNA | JU324403.1 | - | 5 - 6 | 1313 | 70 | |
JU471879.1 | - | 5 - 6 | 1276 | 70 | ||
JU471880.1 | - | 5 - 6 | 1276 | 70 | ||
JV595577.1 | - | 4 - 5 | 1180 | 70 | ||
JV636408.1 | - | 5 - 6 | 1243 | 70 | ||
JV636409.1 | - | 5 - 6 | 1243 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |