Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Oc06726526_g1
          See other LOC103345989 GE Assays ›
          Gene Symbol:
          LOC103345989
          Gene Name:
          Ig heavy chain V-III region 23
          Gene Aliases:
          IGH, IGHVDJ, VH
          Chromosome Location:
          Chr.Unknown: 269006 - 281597 on Build OryCun2.0
          Species:
          Rabbit
          Species Specific ID (Flybase ID):
          -
          Assay ID Oc06726526_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103345989
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017339206.1 XP_017194695.1 109
          GenBank mRNA AF264440.1 - 122 109
          AF264452.1 - 122 109
          AF264465.1 - 122 109
          AF264471.1 - 128 109
          AF264495.1 - 1 128 109
          AF264524.1 - 122 109
          AF264531.1 - 122 109
          AF264538.1 - 122 109
          AF264539.1 - 122 109
          AF264548.1 - 122 109
          AF264550.1 - 122 109
          AF264555.1 - 122 109
          AF264590.1 - 122 109
          AF264593.1 - 122 109
          AF264602.1 - 122 109
          AF264604.1 - 122 109
          AY676695.1 - 1 133 109
          AY676698.1 - 1 133 109
          AY676699.1 - 1 133 109
          AY676700.1 - 1 132 109
          AY676709.1 - 1 133 109
          AY676759.1 - 1 122 109
          AY676761.1 - 1 122 109
          AY676767.1 - 1 122 109
          AY676769.1 - 1 122 109
          AY676770.1 - 1 122 109
          AY676772.1 - 1 122 109
          AY676773.1 - 1 122 109
          AY676774.1 - 1 122 109
          AY676782.1 - 1 131 109
          AY676784.1 - 1 122 109
          AY676786.1 - 1 122 109
          AY676788.1 - 1 122 109
          AY676789.1 - 1 122 109
          AY676790.1 - 1 122 109
          AY676791.1 - 1 122 109
          AY676792.1 - 1 133 109
          AY676793.1 - 1 133 109
          AY676798.1 - 1 122 109
          AY676799.1 - 1 122 109
          AY676804.1 - 1 133 109
          AY676809.1 - 1 133 109
          AY676810.1 - 1 133 109
          AY676812.1 - 1 133 109
          AY676813.1 - 1 133 109
          AY676814.1 - 1 133 109
          AY676819.1 - 1 134 109
          AY676821.1 - 1 133 109
          AY676822.1 - 1 133 109
          AY676831.1 - 1 133 109
          AY676839.1 - 1 133 109
          AY676842.1 - 1 133 109
          AY676843.1 - 1 133 109
          AY676844.1 - 1 132 109
          AY676853.1 - 1 133 109
          L03862.1 - 179 109
          L03897.1 - 179 109
          M77043.2 - 164 109
          M99543.1 - 89 109
          M99544.1 - 89 109
          M99549.1 - 122 109
          M99550.1 - 122 109

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0