Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105607283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976323.1 | XP_027832124.1 | 5 - 6 | 883 | 107 |
Set Membership: |
![]() ![]() ![]() |