Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105607283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976323.1 | XP_027832124.1 | 14 - 15 | 1856 | 57 |
Set Membership: |
![]() ![]() ![]() ![]() |