Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101122753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104525.3 | XP_011959915.3 | 9931 | 100 | ||
XM_027961385.1 | XP_027817186.1 | 9933 | 100 | |||
XM_027961387.1 | XP_027817188.1 | 9669 | 100 | |||
XM_027961388.1 | XP_027817189.1 | 9621 | 100 |