Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101121720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102542.2 | XP_014958028.1 | 5 - 6 | 608 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |