Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04934657_m1
          See other AP5S1 GE Assays ›
          Gene Symbol:
          Ap5s1
          Gene Name:
          adaptor-related protein 5 complex, sigma 1 subunit
          Gene Aliases:
          0610038L13Rik, 2310035K24Rik, AW060727, sigma5
          Chromosome Location:
          Chr.2: 131210360 - 131213514 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04934657_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap5s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110992.1 1 - 2 326 87
          GenBank mRNA BY264259.1 - 1 - 2 327 87
          CX204398.1 - 1 - 2 164 87
          DV061712.1 - 1 - 2 359 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ061061.1 - 1 - 2 262 Chr. 2 - 131210424 - 131212634 [+] on Build GRCm38 87
          GenBankmRNA CX215186.1 - 1 - 2 303 Chr. 2 - 131210383 - 131212715 [+] on Build GRCm38 87
          GenBankmRNA AW743091.1 - 1 - 2 218 Chr. 2 - 131210468 - 131212699 [+] on Build GRCm38 87
          GenBankmRNA DV649720.1 - 1 - 2 303 Chr. 2 - 131210383 - 131212801 [+] on Build GRCm38 87
          GenBankmRNA AA542019.1 - 1 - 2 271 Chr. 2 - 131210413 - 131212742 [+] on Build GRCm38 87
          GenBankmRNA AV449230.1 - 1 - 2 304 Chr. 2 - 131210382 - 131212654 [+] on Build GRCm38 87
          GenBankmRNA BY242492.1 - 1 - 2 318 Chr. 2 - 131210369 - 131212585 [+] on Build GRCm38 87
          GenBankmRNA AI155936.1 - 1 - 2 219 Chr. 2 - 131210467 - 131212650 [+] on Build GRCm38 87
          GenBankmRNA BG086625.2 - 1 - 2 116 Chr. 2 - 131210570 - 131212701 [+] on Build GRCm38 87
          GenBankmRNA CF950821.1 - 1 - 2 252 Chr. 2 - 131210434 - 131212603 [+] on Build GRCm38 87
          GenBankmRNA CF737427.1 - 1 - 2 266 Chr. 2 - 131210420 - 131212840 [+] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cell division cycle 25B Cdc25b AI604853 -11.8kb Sense Chr. 2 - 131186436 - 131198511 on NCBI Build GRCm38
          Downstream mitochondrial antiviral signaling protein Mavs D430028G21Rik,
          IPS-1,
          Visa,
          cardif
          20.5kb Sense Chr. 2 - 131234063 - 131248025 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA repair
          transport
          cellular response to DNA damage stimulus
          protein transport
          endosomal transport
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0