Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04410435_m1
          See other C430002N11RIK GE Assays ›
          Gene Symbol:
          C430002N11Rik
          Gene Name:
          RIKEN cDNA C430002N11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 96765562 - 96774397 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04410435_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C430002N11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102293.1 4 - 5 430 65
          NR_102296.1 4 - 5 517 65
          GenBank mRNA AA030139.1 - 4 - 5 391 65
          AK049386.1 - 4 - 5 505 65
          AK089981.1 - 504 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA E030011O05 gene E030011O05Rik -18.7kb Anti-sense Chr. 9 - 96731575 - 96752831 on NCBI Build GRCm38
          Downstream 2-phosphoxylose phosphatase 1 Pxylp1 9430094M07Rik,
          Acpl2,
          BB177120,
          C130099A20Rik
          48.9kb Anti-sense Chr. 9 - 96823336 - 96892680 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001218.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2427150 AK049386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2427150 BB407702.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118547 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135467 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2427150 AK049386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2427150 BB407702.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02006245 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135467 - -
          Whole Mouse Genome G4122A A_52_P239470 AK049386.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115327 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08116735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07839967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07543508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07947639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07839968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07870876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07870877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08116739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08188632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26748964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440510_PM_at - -
          Mouse Genome 430 2.0 Array 1440510_at - -
          Mouse Genome 430 2.0 Array 1440510_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900009587.mm.1 TC0900001218.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009587.mm.1 NR_102293.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009587.mm.1 NR_102296.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009588.mm.1 TC0900001218.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009588.mm.1 NR_102293.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009588.mm.1 NR_102294.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009588.mm.1 NR_102295.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009588.mm.1 NR_102296.1 Refseq
          Mouse Expression Set 430 1440510_at - -
          Mouse Expression Set 430 1440510_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440510_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440510_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757853 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102810097 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0