Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336276_s1
          See other 4930483O08RIK GE Assays ›
          Gene Symbol:
          4930483O08Rik
          Gene Name:
          RIKEN cDNA 4930483O08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 133128251 - 133142457 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336276_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930483O08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046279.1 3 717 76
          GenBank mRNA AK015618.1 - 3 718 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream C-terminal binding protein 2 Ctbp2 AA407280,
          D7Ertd45e,
          Gtrgeo6,
          Ribeye
          -17.9kb Anti-sense Chr. 7 - 132986417 - 133123877 on NCBI Build GRCm38
          Downstream testis expressed 36 Tex36 4930404H21Rik 444.6kb Anti-sense Chr. 7 - 133587024 - 133602115 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001945.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0100002760.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P06018899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06654906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05986264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06340830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05924968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25539098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06174146 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454208_PM_at - -
          Mouse Genome 430 2.0 Array 1454208_PM_at - -
          Mouse Genome 430 2.0 Array 1454208_at - -
          Affymetrix Mouse Clariom S Array PSR0700017023.mm.1 TC0700001945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017023.mm.1 NR_046279.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017023.mm.1 NR_046279.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017022.mm.1 TC0700001945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017022.mm.1 NR_046279.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017022.mm.1 NR_046279.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017021.mm.1 TC0700001945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017021.mm.1 NR_046279.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017021.mm.1 NR_046279.1 Refseq
          Mouse Expression Set 430 1454208_PM_at - -
          Mouse Expression Set 430 1454208_at - -
          Affymetrix HT Mouse Genome 430B Array 1454208_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454208_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 791989 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102640079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0