Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336269_s1
          See other 9130221F21RIK GE Assays ›
          Gene Symbol:
          9130221F21Rik
          Gene Name:
          RIKEN cDNA 9130221F21 gene
          Gene Aliases:
          9130221F21
          Chromosome Location:
          Chr.6: 72013896 - 72042528 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336269_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9130221F21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046180.1 3 1724 89
          GenBank mRNA AK033688.1 - 3 1725 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream polymerase (RNA) I polypeptide A Polr1a 194kDa,
          3010014K16Rik,
          RPA194,
          Rpo1-4,
          mRPA1
          -34.5kb Anti-sense Chr. 6 - 71909053 - 71979435 on NCBI Build GRCm38
          Downstream ST3 beta-galactoside alpha-2,3-sialyltransferase 5 St3gal5 3S-T,
          Siat9,
          [a]2
          81.6kb Anti-sense Chr. 6 - 72097554 - 72154570 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002477.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675810 AK020236.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176766 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150835 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150835 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100246 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05176304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24539651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600019179.mm.1 TC0600002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019179.mm.1 NR_046180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019179.mm.1 NR_046180.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019177.mm.1 TC0600002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019177.mm.1 NR_046180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019177.mm.1 NR_046180.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019178.mm.1 TC0600002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019178.mm.1 NR_046180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019178.mm.1 NR_046180.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0