Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336057_m1
          See other AI506816 GE Assays ›
          Gene Symbol:
          AI506816
          Gene Name:
          expressed sequence AI506816
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 23692261 - 23712667 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336057_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI506816
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038090.1 2 - 3 63 101
          GenBank mRNA CJ127083.1 - 2 - 3 63 101
          CJ128682.1 - 2 - 3 63 101
          CJ130089.1 - 2 - 3 58 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ129998.1 - 1 - 2 51 Chr. 5 - 23710039 - 23711153 [-] on Build GRCm38 101
          GenBankmRNA CJ129220.1 - 2 - 3 63 Chr. 5 - 23710059 - 23712667 [-] on Build GRCm38 101
          GenBankmRNA CJ130468.1 - 2 - 3 58 Chr. 5 - 23710018 - 23712662 [-] on Build GRCm38 101
          GenBankmRNA CJ125853.1 - 2 - 3 63 Chr. 5 - 23710022 - 23712667 [-] on Build GRCm38 101
          GenBankmRNA CJ103915.1 - 1 - 2 49 Chr. 5 - 23710090 - 23711153 [-] on Build GRCm38 101
          GenBankmRNA CJ130151.1 - 2 - 3 58 Chr. 5 - 23710061 - 23712662 [-] on Build GRCm38 101
          GenBankmRNA CJ129028.1 - 2 - 3 63 Chr. 5 - 23710036 - 23712667 [-] on Build GRCm38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream serine/arginine-rich protein specific kinase 2 Srpk2 AW226533,
          AW492537,
          AW547358,
          Wbp6,
          mSRPK2
          -25.4kb Sense Chr. 5 - 23503356 - 23684641 on NCBI Build GRCm38
          Downstream pseudouridylate synthase 7 homolog (S. cerevisiae) Pus7 C330017I15Rik 29.0kb Sense Chr. 5 - 23740165 - 23783711 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002046.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23538005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04976954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04658503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04976956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04494327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04753164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23538001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23538006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23538002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04127724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23537932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351506 - -
          Affymetrix HT MG-430 PM Array Plate 1439483_PM_at - -
          Mouse Genome 430 2.0 Array 1439483_at - -
          Mouse Genome 430 2.0 Array 1439483_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500018648.mm.1 TC0500002046.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018648.mm.1 NR_015554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018648.mm.1 NR_015554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018648.mm.1 NR_038090.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018648.mm.1 NR_038090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018640.mm.1 TC0500002046.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018640.mm.1 NR_038090.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018640.mm.1 NR_038090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018642.mm.1 TC0500002046.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018642.mm.1 NR_015554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018642.mm.1 NR_015554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018642.mm.1 NR_038090.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018642.mm.1 NR_038090.1 Refseq
          Mouse Expression Set 430 1439483_PM_at - -
          Mouse Expression Set 430 1439483_at - -
          Murine Genome U74v2 Set 97197_PM_r_at - -
          Murine Genome U74v2 Set 97197_r_at - -
          Mouse Genome 430A 2.0 Array 1439483_PM_at - -
          Mouse Genome 430A 2.0 Array 1439483_at - -
          Affymetrix HT Mouse Genome 430A Array 1439483_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439483_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0