Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335952_m1
          See other GM3279 GE Assays ›
          Gene Symbol:
          Gm3279
          Gene Name:
          predicted gene 3279
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 55684571 - 55694574 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335952_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm3279
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046071.1 1 - 2 41 80
          GenBank mRNA AK038589.1 - 1 - 2 41 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB631873.1 - 1 - 2 41 Chr. 6 - 55684571 - 55694116 [+] on Build GRCm38 80
          GenBankmRNA BB631901.1 - 1 - 2 41 Chr. 6 - 55684571 - 55694138 [+] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream neurogenic differentiation 6 Neurod6 Atoh2,
          Math-2,
          Math2,
          Nex,
          Nex1m,
          bHLHa2
          -3.3kb Anti-sense Chr. 6 - 55677818 - 55681263 on NCBI Build GRCm38
          Downstream coiled-coil domain containing 129 Ccdc129 D530004J12Rik,
          Gm458
          116.4kb Sense Chr. 6 - 55804628 - 55978743 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000669.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691581 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139410 AK038589.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01335635 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139410 AK038589.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05645006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05781561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05644998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05644999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05234342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05792650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05645001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24471164 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600005189.mm.1 TC0600000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005189.mm.1 NR_046071.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005189.mm.1 NR_046071.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005190.mm.1 TC0600000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005190.mm.1 NR_046071.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005190.mm.1 NR_046071.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005191.mm.1 TC0600000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005191.mm.1 NR_046071.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005191.mm.1 NR_046071.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 845165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0