Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335943_m1
          See other GM11240 GE Assays ›
          Gene Symbol:
          Gm11240
          Gene Name:
          predicted gene 11240
          Gene Aliases:
          OTTMUSG00000000286
          Chromosome Location:
          Chr.4: 73791244 - 73798378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335943_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11240
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046041.1 2 - 3 333 72
          NR_046042.1 2 - 3 262 72
          GenBank mRNA AK131821.1 - 2 - 3 335 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707593.1 - 2 - 3 331 Chr. 4 - 73791246 - 73798352 [+] on Build GRCm38 72
          GenBankmRNA CF616660.1 - 2 - 3 259 Chr. 4 - 73791248 - 73798380 [+] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RAS and EF hand domain containing Rasef BC030882,
          RAB45
          -2.5kb Anti-sense Chr. 4 - 73714577 - 73791401 on NCBI Build GRCm38
          Downstream predicted gene 11758 Gm11758 OTTMUSG00000004013 73.8kb Anti-sense Chr. 4 - 73872219 - 73876266 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000772.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115539 AK131821.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 AK034935.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 AK043681.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 AK084021.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 BC116730.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 BC117087.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841430 NM_175564.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138062 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160197 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115539 AK131821.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160197 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400005446.mm.1 TC0400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005446.mm.1 NR_046041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005446.mm.1 NR_046041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005446.mm.1 NR_046042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005446.mm.1 NR_046042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005445.mm.1 TC0400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005445.mm.1 NR_046041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005445.mm.1 NR_046041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005445.mm.1 NR_046042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005445.mm.1 NR_046042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005447.mm.1 TC0400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005447.mm.1 NR_046041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005447.mm.1 NR_046041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005447.mm.1 NR_046042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005447.mm.1 NR_046042.1 Refseq
          Murine Genome U74v2 Set 167027_PM_at - -
          Murine Genome U74v2 Set 167027_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0