Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335935_m1
          See other GM16938 GE Assays ›
          Gene Symbol:
          Gm16938
          Gene Name:
          predicted gene, 16938
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 98177194 - 98184804 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335935_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16938
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045970.1 2 - 3 406 84
          NR_045971.1 2 - 3 406 84
          GenBank mRNA BC144933.1 - 2 - 3 359 84
          BC144936.1 - 2 - 3 359 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream olfactory marker protein Omp -33.0kb Anti-sense Chr. 7 - 98143359 - 98145447 on NCBI Build GRCm38
          Downstream UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase) B3gnt6 BC039789,
          Core-3,
          Core3
          13.0kb Anti-sense Chr. 7 - 98192415 - 98199475 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001364.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131046 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01585143 - -
          Whole Mouse Genome G4122A A_52_P318426 AK085722.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 TC0700001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045972.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011612.mm.1 NR_045972.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 TC0700001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045972.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011609.mm.1 NR_045972.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 TC0700001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045972.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011613.mm.1 NR_045972.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 888699 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101940538 - -
          MouseWG-6 v2 Expression BeadChip 2570114 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-69f987f899-9tng4:80/100.66.130.250:80.
          git-commit: eb1b8c2b8eec225754209f035b16fef014668c1b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0