Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335911_m1
          See other GM10791 GE Assays ›
          Gene Symbol:
          Gm10791
          Gene Name:
          predicted gene 10791
          Gene Aliases:
          ENSMUSG00000074933
          Chromosome Location:
          Chr.16: 84972211 - 84979451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335911_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10791
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045889.1 1 - 2 541 53
          GenBank mRNA AK166404.1 - 1 - 2 123 53
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY735116.1 - 1 - 2 123 Chr. 16 - 84972630 - 84978876 [+] on Build GRCm38 53

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream GA repeat binding protein,
          alpha
          Gabpa GABPalpha -108.4kb Sense Chr. 16 - 84834925 - 84863783 on NCBI Build GRCm38
          Downstream cysteine and tyrosine-rich protein 1 Cyyr1 476.6kb Anti-sense Chr. 16 - 85450357 - 85550905 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000941.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK166404.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994289 AK166404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994290 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994289 AK166404.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1447012_PM_at - -
          Mouse Genome 430 2.0 Array 1447012_PM_at - -
          Mouse Genome 430 2.0 Array 1447012_at - -
          Affymetrix Mouse Clariom S Array PSR1600007372.mm.1 TC1600000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007372.mm.1 NR_045889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007372.mm.1 NR_045889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007374.mm.1 TC1600000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007374.mm.1 NR_045889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007374.mm.1 NR_045889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007375.mm.1 TC1600000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007375.mm.1 NR_045889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007375.mm.1 NR_045889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007377.mm.1 TC1600000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007377.mm.1 NR_045889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007377.mm.1 NR_045889.1 Refseq
          Mouse Expression Set 430 1447012_PM_at - -
          Mouse Expression Set 430 1447012_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 516503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0