Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045885.1 | 3 - 4 | 407 | 80 | ||
GenBank mRNA | AK133282.1 | - | 3 - 4 | 408 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | O-6-methylguanine-DNA methyltransferase | Mgmt |
AGT, AI267024, Agat |
-110.1kb | Sense | Chr. 7 - 136894468 - 137128193 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 9430038I01 gene | 9430038I01Rik | 133.0kb | Anti-sense | Chr. 7 - 137375569 - 137411900 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() |