Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04279164_s1
          See other 9330117O12RIK GE Assays ›
          Gene Symbol:
          9330117O12Rik
          Gene Name:
          RIKEN cDNA 9330117O12 gene
          Gene Aliases:
          9330117O12
          Chromosome Location:
          Chr.18: 54611744 - 54639870 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04279164_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330117O12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045400.1 3 1620 122
          GenBank mRNA AK033928.1 - 3 1621 122
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB076153.2 - 1 436 Chr. 18 - 54639215 - 54639866 [+] on Build GRCm38 122

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Redrum,
          erythroid developmental long intergenic non-protein coding transcript
          Redrum 9430076G02Rik -185.3kb Sense Chr. 18 - 54422295 - 54453294 on NCBI Build GRCm38
          Downstream predicted gene,
          33732
          Gm33732 162.7kb Anti-sense Chr. 18 - 54802618 - 54840255 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09006201 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13945563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13985264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13620965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13721770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31719909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13721771 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 539469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0