Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04279089_s1
          See other 7630403G23RIK GE Assays ›
          Gene Symbol:
          7630403G23Rik
          Gene Name:
          RIKEN cDNA 7630403G23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 34042371 - 34059019 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04279089_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 7630403G23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040744.1 1 423 104
          GenBank mRNA AK020190.1 - 1 425 104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY720400.1 - 1 423 Chr. 9 - 34042371 - 34058630 [+] on Build GRCm38 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 27166 Gm27166 -951.6kb Sense Chr. 9 - 33031791 - 33090799 on NCBI Build GRCm38
          Downstream kin of IRRE like 3 (Drosophila) Kirrel3 1500010O20Rik,
          2900036G11Rik,
          NEPH2,
          SST4,
          mKIAA1867
          443.0kb Sense Chr. 9 - 34485852 - 35036315 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000311.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0800000354.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2409038 AK020190.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1436 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01902593 - -
          Whole Mouse Genome G4122A A_52_P730528 AK020190.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07670715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07460897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08053046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07870021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07499533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07840977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07819884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07794912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07819895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07499536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08144987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26414206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430881_PM_at - -
          Mouse Genome 430 2.0 Array 1430881_at - -
          Mouse Genome 430 2.0 Array 1430881_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900002322.mm.1 TC0900000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002322.mm.1 NR_040744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002322.mm.1 NR_040744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002320.mm.1 TC0900000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002320.mm.1 NR_040744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002320.mm.1 NR_040744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002321.mm.1 TC0900000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002321.mm.1 NR_040744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002321.mm.1 NR_040744.1 Refseq
          Mouse Expression Set 430 1430881_PM_at - -
          Mouse Expression Set 430 1430881_at - -
          Affymetrix HT Mouse Genome 430B Array 1430881_at - -
          Affymetrix HT Mouse Genome 430B Array 1430881_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 840775 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0