Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700120E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045368.1 | 3 - 4 | 1496 | 96 | ||
GenBank mRNA | AK007214.1 | - | 3 - 4 | 1497 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myosin VB | Myo5b |
AI661750, mKIAA1119 |
Anti-sense | Chr. 18 - 74442378 - 74772698 on NCBI Build GRCm38 | |
Downstream | small Cajal body-specific RNA 17 | Scarna17 | MBII-119 | 255.1kb | Sense | Chr. 18 - 74778394 - 74778455 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |