Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278361_m1
          See other 4930547E14RIK GE Assays ›
          Gene Symbol:
          4930547E14Rik
          Gene Name:
          RIKEN cDNA 4930547E14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 59636945 - 59672993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278361_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930547E14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040564.1 1 - 2 238 120
          GenBank mRNA AK016054.1 - 1 - 2 239 120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715653.1 - 1 - 2 238 Chr. 16 - 59636945 - 59672995 [+] on Build GRCm38 120

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ADP-ribosylation factor-like 6 Arl6 1110018H24Rik,
          2210411E14Rik,
          BBS3
          Anti-sense Chr. 16 - 59613321 - 59639405 on NCBI Build GRCm38
          Downstream NOL1/NOP2/Sun domain family member 3 Nsun3 6720484A09Rik,
          AU022521
          3.1mb Anti-sense Chr. 16 - 62732444 - 62786794 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000774.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34173 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08614976 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122688 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600006719.mm.1 TC1600000774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006719.mm.1 NR_040564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006719.mm.1 NR_040564.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006714.mm.1 TC1600000774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006714.mm.1 NR_040564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006714.mm.1 NR_040564.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006721.mm.1 TC1600000774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006721.mm.1 NR_040564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006721.mm.1 NR_040564.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580748 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0