Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278357_m1
          See other 1700112H15RIK GE Assays ›
          Gene Symbol:
          1700112H15Rik
          Gene Name:
          RIKEN cDNA 1700112H15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 184527841 - 184557691 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278357_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700112H15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040472.1 4 - 5 313 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dual specificity phosphatase 10 Dusp10 2610306G15Rik,
          AI158871,
          MKP-5,
          MKP5
          -453.2kb Anti-sense Chr. 1 - 184034461 - 184075636 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930488B22 gene 4930488B22Rik 182.3kb Anti-sense Chr. 1 - 184712497 - 184718989 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003734.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105717 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04383579 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04383599 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04383636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04383599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00279633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00903674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00568166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00568169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00903669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00092354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00568164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00730530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01007395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00730531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00376917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20895972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20896040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100030587.mm.1 TC0100003734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100030587.mm.1 NR_040472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100030587.mm.1 NR_040472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100030588.mm.1 TC0100003734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100030588.mm.1 NR_040472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100030588.mm.1 NR_040472.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-785d8696b7-btkzg:80/100.66.135.120:80.
          git-commit: 410fd2c6c8b73ec5a2f927f842de1c3d37a6dc6e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0