Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040305.1 | 2 - 3 | 656 | 69 | ||
GenBank mRNA | AK138296.1 | - | 2 - 3 | 657 | 69 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 9630028B13 gene | 9630028B13Rik | -5.3kb | Sense | Chr. 1 - 185429357 - 185441819 on NCBI Build GRCm38 | |
Downstream |
solute carrier family 30, member 10 |
Slc30a10 |
E130106K10Rik, Gm212 |
715.0 | Anti-sense | Chr. 1 - 185454848 - 185468762 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() |