Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1372-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034155.1 | 2 | 947 | 80 | ||
GenBank mRNA | AK137189.1 | - | 1886 | 80 | ||
BC055827.1 | - | 2 | 950 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactory receptor 1373 | Olfr1373 | GA_x6K02T2QP88-3283154-3284089 | -9.3kb | Sense | Chr. 11 - 52144593 - 52145528 on NCBI Build GRCm38 |
Downstream | olfactory receptor 1371 | Olfr1371 |
GA_x6K02T2QP88-3214035-3214970, MOR256-58 |
56.5kb | Sense | Chr. 11 - 52213050 - 52224497 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |