Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242154_m1
          See other GM7056 GE Assays ›
          Gene Symbol:
          Gm7056
          Gene Name:
          predicted gene 7056
          Gene Aliases:
          Gm149
          Chromosome Location:
          Chr.5: 89616436 - 89622611 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242154_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm7056
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037571.1 140 97
          GenBank mRNA AK135800.1 - 1 - 2 130 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF915070.1 - 1 - 2 138 Chr. 5 - 89616438 - 89620126 [+] on Build GRCm38 97
          GenBankmRNA CF172806.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620269 [+] on Build GRCm38 97
          GenBankmRNA CF916606.1 - 1 - 2 112 Chr. 5 - 89616464 - 89619874 [+] on Build GRCm38 97
          GenBankmRNA CF914285.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620149 [+] on Build GRCm38 97
          GenBankmRNA CA559515.1 - 1 - 2 108 Chr. 5 - 89616468 - 89620094 [+] on Build GRCm38 97
          GenBankmRNA CF174411.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620434 [+] on Build GRCm38 97
          GenBankmRNA DV654294.1 - 1 - 2 125 Chr. 5 - 89616451 - 89619853 [+] on Build GRCm38 97
          GenBankmRNA CF172058.1 - 1 - 2 108 Chr. 5 - 89616468 - 89620168 [+] on Build GRCm38 97
          GenBankmRNA CA556045.1 - 1 - 2 104 Chr. 5 - 89616472 - 89619775 [+] on Build GRCm38 97
          GenBankmRNA CF913793.1 - 1 - 2 138 Chr. 5 - 89616438 - 89620083 [+] on Build GRCm38 97
          GenBankmRNA CF174503.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620161 [+] on Build GRCm38 97
          GenBankmRNA CF172642.1 - 1 - 2 104 Chr. 5 - 89616472 - 89620211 [+] on Build GRCm38 97
          GenBankmRNA CF174166.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620116 [+] on Build GRCm38 97
          GenBankmRNA CA562173.1 - 1 - 2 104 Chr. 5 - 89616472 - 89620006 [+] on Build GRCm38 97
          GenBankmRNA CF174703.1 - 1 - 2 140 Chr. 5 - 89616436 - 89620115 [+] on Build GRCm38 97
          GenBankmRNA CJ052630.1 - 1 - 2 130 Chr. 5 - 89616447 - 89620160 [+] on Build GRCm38 97
          GenBankmRNA CA560393.1 - 1 - 2 124 Chr. 5 - 89616452 - 89620034 [+] on Build GRCm38 97
          GenBankmRNA CF914282.1 - 1 - 2 108 Chr. 5 - 89616468 - 89620185 [+] on Build GRCm38 97
          GenBankmRNA CF173226.1 - 1 - 2 138 Chr. 5 - 89616438 - 89620161 [+] on Build GRCm38 97
          GenBankmRNA CF914574.1 - 1 - 2 104 Chr. 5 - 89616472 - 89620136 [+] on Build GRCm38 97
          GenBankmRNA CF172456.1 - 1 - 2 108 Chr. 5 - 89616468 - 89620195 [+] on Build GRCm38 97
          GenBankmRNA CF916049.1 - 1 - 2 112 Chr. 5 - 89616464 - 89620137 [+] on Build GRCm38 97
          GenBankmRNA CA558047.1 - 1 - 2 138 Chr. 5 - 89616438 - 89620031 [+] on Build GRCm38 97
          GenBankmRNA CA560141.1 - 1 - 2 108 Chr. 5 - 89616468 - 89622056 [+] on Build GRCm38 97
          GenBankmRNA CA558593.1 - 1 - 2 140 Chr. 5 - 89616436 - 89620084 [+] on Build GRCm38 97
          GenBankmRNA CF172053.1 - 1 - 2 141 Chr. 5 - 89616436 - 89620149 [+] on Build GRCm38 97
          GenBankmRNA CF914286.1 - 1 - 2 118 Chr. 5 - 89616464 - 89620095 [+] on Build GRCm38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream neuropeptide FF receptor 2 Npffr2 Gpr74,
          HG31,
          NPFF2
          -32.6kb Sense Chr. 5 - 89526577 - 89583914 on NCBI Build GRCm38
          Downstream a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif,
          3
          Adamts3 1100001H14Rik,
          6330442E02Rik
          53.9kb Anti-sense Chr. 5 - 89673841 - 89883669 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000828.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P188856 NR_037571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P188856 NR_037571.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420490 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P188856 NR_037571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P188856 NR_037571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188856 NR_037571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188856 NR_037571.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P04531250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04107216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04516731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04531248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04738289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04391654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04684343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04165488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04531251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04287759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04437704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04391634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04684346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23878626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04287763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982738 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 631105_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 631105_at - -
          Affymetrix Mouse Clariom S Array PSR0500007308.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007308.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007308.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007305.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007305.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007305.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007310.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007310.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007310.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007307.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007307.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007307.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007302.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007302.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007302.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007304.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007304.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007304.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007309.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007309.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007309.mm.1 NR_037571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007306.mm.1 TC0500000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007306.mm.1 NR_037571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007306.mm.1 NR_037571.1 Refseq
          Mouse Expression Set 430 1437178_PM_at - -
          Mouse Expression Set 430 1437178_at - -
          Murine Genome U74v2 Set 131074_at - -
          Murine Genome U74v2 Set 131074_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428754 - -
          ABI Mouse Genome Survey Microarray 901269 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588071 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-hbrf6:80/100.66.134.79:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0