Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04240921_m1
          See other GM5084 GE Assays ›
          Gene Symbol:
          Gm5084
          Gene Name:
          predicted gene 5084
          Gene Aliases:
          A030007L22, EG328264
          Chromosome Location:
          Chr.13: 60205471 - 60216018 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04240921_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5084
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036449.1 3 - 4 2858 82
          GenBank mRNA AK037190.1 - 3 - 4 2859 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX515698.1 - 1 - 2 226 Chr. 13 - 60212915 - 60215580 [+] on Build GRCm38 82
          GenBankmRNA AI464751.1 - 1 - 2 225 Chr. 13 - 60212916 - 60215582 [+] on Build GRCm38 82
          GenBankmRNA AA255008.1 - 1 - 2 188 Chr. 13 - 60212953 - 60215459 [+] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream growth arrest specific 1 Gas1 AW554192,
          Gas-1
          -34.3kb Anti-sense Chr. 13 - 60174405 - 60177535 on NCBI Build GRCm38
          Downstream RIKEN cDNA A530001N23 gene A530001N23Rik 267.9kb Anti-sense Chr. 13 - 60483921 - 60508840 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000731.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557395 AK037190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557395 NR_036449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557395 AK037190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557395 NR_036449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557395 AK037190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557395 NR_036449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557395 AK037190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557395 NR_036449.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10588310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10548498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10548497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10438332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10689859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11043188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10549385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29084110 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300004864.mm.1 TC1300000731.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004864.mm.1 NR_036449.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004864.mm.1 NR_036449.1 Refseq
          Mouse Expression Set 430 1459259_at - -
          Mouse Expression Set 430 1459259_PM_at - -
          Murine Genome U74v2 Set 134808_at - -
          Murine Genome U74v2 Set 134808_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fbcb5ff59-4c8rf:80/100.66.130.250:80.
          git-commit: f911e8b0a61717f033c2f2c0e0d8be5576cef2f2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0