Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239588_m1
          See other FMR1OS GE Assays ›
          Gene Symbol:
          Fmr1os
          Gene Name:
          fragile X mental retardation syndrome 1, opposite strand
          Gene Aliases:
          Asfmr1, ENSMUSG00000073146, Gm10474
          Chromosome Location:
          Chr.X: 68667514 - 68678399 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239588_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fmr1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033481.1 3 - 4 390 128
          GenBank mRNA AK148387.1 - 3 - 4 391 128
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY741516.1 - 3 - 4 388 Chr. X - 68669523 - 68678397 [-] on Build GRCm38 128

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 509 Mir509 Mirn509,
          mmu-mir-509
          -657.3kb Sense Chr. X - 68010105 - 68010179 on NCBI Build GRCm38
          Downstream fragile X mental retardation syndrome 1 Fmr1 FMRP,
          Fmr-1
          5.2kb Anti-sense Chr. X - 68678541 - 68717963 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002373.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK148387.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053004 AK148387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053004 NR_033481.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119658 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09319508 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09319508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32440999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14517296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14627910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32441028 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10604895 - -
          Affymetrix Mouse Clariom S Array PSR0X00014705.mm.1 TC0X00002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00014705.mm.1 NR_033481.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014705.mm.1 NR_033481.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0