Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04236103_m1
          See other MUG-PS1 GE Assays ›
          Gene Symbol:
          Mug-ps1
          Gene Name:
          murinoglobulin, pseudogene 1
          Gene Aliases:
          Mug3
          Chromosome Location:
          Chr.6: 122176053 - 122248566 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04236103_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mug-ps1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027619.1 17 - 18 2188 115
          GenBank mRNA M65237.1 - 4 - 5 801 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream murinoglobulin pseudogene Gm10319 EG381806,
          ENSMUSG00000071204
          -81.8kb Sense Chr. 6 - 122136628 - 122150958 on NCBI Build GRCm38
          Downstream killer cell lectin-like receptor subfamily G,
          member 1
          Klrg1 2F1-Ag,
          MAFA,
          MAFA-L
          36.7kb Anti-sense Chr. 6 - 122270596 - 122282833 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001376.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170379 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06051720 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06051742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05453824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05159053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05453832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05453823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541451 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541480 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541448 - -
          Affymetrix Mouse Clariom S Array PSR0600011114.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011114.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011121.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011121.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011105.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011105.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011108.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011108.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011118.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011118.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011110.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011110.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011112.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011112.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011117.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011117.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011103.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011103.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011104.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011104.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011111.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011111.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011106.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011106.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011107.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011107.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011109.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011109.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011119.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011119.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011116.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011116.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011120.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011120.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011113.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011113.mm.1 NR_027619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011115.mm.1 TC0600001376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011115.mm.1 NR_027619.1 Refseq
          Mouse Expression Set 430 1448854_PM_s_at - -
          Mouse Expression Set 430 1448854_s_at - -
          Murine Genome U74v2 Set 97680_PM_at - -
          Murine Genome U74v2 Set 97680_at - -
          Mouse Genome 430A 2.0 Array 1448854_s_at - -
          Mouse Genome 430A 2.0 Array 1448854_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770138 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
          git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0