Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235551_g1
          See other DANCR GE Assays ›
          Gene Symbol:
          Dancr
          Gene Name:
          differentiation antagonizing non-protein coding RNA
          Gene Aliases:
          2700023E23Rik
          Chromosome Location:
          Chr.5: 74093083 - 74094336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235551_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dancr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015531.1 1 - 2 61 100
          GenBank mRNA AK172090.1 - 1 - 2 61 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY215235.1 - 1 - 2 61 Chr. 5 - 74093083 - 74093644 [+] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 26
          Snora26 MBI-6 Sense Chr. 5 - 74093530 - 74093650 on NCBI Build GRCm38
          Downstream RAS-like,
          family 11,
          member B
          Rasl11b 1190017B18Rik 101.0kb Sense Chr. 5 - 74195326 - 74199477 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000697.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095118 AK045300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095118 AK172090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095118 AK172703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095118 NR_015531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095118 AK012278.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104092 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095118 AK045300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095118 AK172090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095118 AK172703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095118 NR_015531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095118 AK012278.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104092 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122943 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04202079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04346194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1455878_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522465 - -
          Mouse Genome 430 2.0 Array 1455878_at - -
          Mouse Genome 430 2.0 Array 1455878_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70036_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70036_at - -
          Affymetrix Mouse Clariom S Array PSR0500006159.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006159.mm.1 NR_015531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006159.mm.1 NR_031758.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006159.mm.1 NR_031758.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006160.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006160.mm.1 NR_015531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006161.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006161.mm.1 NR_015531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006155.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006155.mm.1 NR_015531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006158.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006158.mm.1 NR_015531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006157.mm.1 TC0500000697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006157.mm.1 NR_015531.2 Refseq
          Mouse Expression Set 430 1455878_at - -
          Mouse Expression Set 430 1455878_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455878_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455878_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104070594 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230037 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0