Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235544_g1
          See other 2810429I04RIK GE Assays ›
          Gene Symbol:
          2810429I04Rik
          Gene Name:
          RIKEN cDNA 2810429I04 gene
          Gene Aliases:
          AI256682
          Chromosome Location:
          Chr.13: 3478303 - 3492398 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235544_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2810429I04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015522.1 2 - 3 103 98
          GenBank mRNA AK043278.1 - 2 - 3 104 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB257552.2 - 2 - 3 113 Chr. 13 - 3478304 - 3492380 [+] on Build GRCm38 98
          GenBankmRNA BU147524.1 - 2 - 3 129 Chr. 13 - 3478277 - 3485737 [+] on Build GRCm38 98
          GenBankmRNA BB640710.1 - 2 - 3 104 Chr. 13 - 3478303 - 3492406 [+] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream spermatogenesis associated glutamate (E)-rich protein 6,
          pseudogene 1
          Speer6-ps1 1700039A02Rik,
          SPEER-6
          -289.8kb Sense Chr. 13 - 3149475 - 3189678 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700024F13 gene 1700024F13Rik 18.0kb Sense Chr. 13 - 3498050 - 3501393 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000005.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK043092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK043278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK034854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK084238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 NR_015522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK043092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK043278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK034854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 AK084238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P502226 NR_015522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2370219 AK034854.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154600 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59025 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139574 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07770468 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07770446 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139574 - -
          Whole Mouse Genome G4122A A_52_P397909 AK087718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P397909 AK034854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P397909 AK084238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457123 AK087718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457123 AK034854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK043092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK043278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK034854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK084238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 NR_015522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK043092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK043278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK034854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 AK084238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502226 NR_015522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378827 AK031301.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10481028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10901069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10901072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10816666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10863754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28788278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454147_PM_at - -
          Mouse Genome 430 2.0 Array 1454147_PM_at - -
          Mouse Genome 430 2.0 Array 1454147_at - -
          Affymetrix Mouse Clariom S Array PSR1300000029.mm.1 TC1300000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000029.mm.1 NR_015522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000029.mm.1 NR_015522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000030.mm.1 TC1300000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000030.mm.1 NR_015522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000030.mm.1 NR_015522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000037.mm.1 TC1300000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000037.mm.1 NR_015522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000037.mm.1 NR_015522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000028.mm.1 TC1300000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000028.mm.1 NR_015522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000028.mm.1 NR_015522.1 Refseq
          Mouse Expression Set 430 1454147_PM_at - -
          Mouse Expression Set 430 1454147_at - -
          Affymetrix HT Mouse Genome 430B Array 1454147_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454147_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0