Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235541_g1
          See other AI662270 GE Assays ›
          Gene Symbol:
          AI662270
          Gene Name:
          expressed sequence AI662270
          Gene Aliases:
          AW496474
          Chromosome Location:
          Chr.11: 83223576 - 83226584 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235541_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI662270
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015519.1 2 - 3 181 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY400360.1 - 2 - 3 183 Chr. 11 - 83223576 - 83224540 [+] on Build GRCm38 86
          GenBankmRNA AI608069.1 - 1 - 2 163 Chr. 11 - 83224152 - 83224593 [+] on Build GRCm38 86
          GenBankmRNA AI661226.1 - 1 - 2 Chr. 11 - 83224200 - 83224593 [-] on Build GRCm38 86
          GenBankmRNA BY089652.1 - 2 - 3 163 Chr. 11 - 83223594 - 83225575 [+] on Build GRCm38 86
          GenBankmRNA AA254039.1 - 1 - 2 176 Chr. 11 - 83224149 - 83224593 [+] on Build GRCm38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream schlafen 3 Slfn3 -5.8kb Sense Chr. 11 - 83191301 - 83218427 on NCBI Build GRCm38
          Downstream schlafen 14 Slfn14 Gm20,
          Slfn14-ps
          50.5kb Anti-sense Chr. 11 - 83275110 - 83286726 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001243.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078735 AK079857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078735 AK136560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078735 NR_015519.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078735 AK028715.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315975 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176388 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078735 AK079857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078735 AK136560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078735 NR_015519.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078735 AK028715.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176388 - -
          Whole Mouse Genome G4122A A_52_P137193 AK028715.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09638890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09725053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09638891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09148915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970347 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434067_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434068_PM_s_at - -
          Affymetrix Mouse Clariom S Array PSR1100011667.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011667.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011667.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011669.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011669.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011669.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011658.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011658.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011658.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011661.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011661.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011661.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011664.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011664.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011664.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011665.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011665.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011665.mm.1 NR_015519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011668.mm.1 TC1100001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011668.mm.1 NR_015519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011668.mm.1 NR_015519.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 425026 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104200685 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
          git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0