Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235474_m1
          See other 2900041M22RIK GE Assays ›
          Gene Symbol:
          2900041M22Rik
          Gene Name:
          RIKEN cDNA 2900041M22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 117611247 - 117613417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235474_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2900041M22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015489.2 1 - 2 186 70
          GenBank mRNA AK019332.1 - 1 - 2 59 70
          AK038849.1 - 1 - 2 90 70
          AK038850.1 - 1 - 2 90 70
          AK046435.1 - 1 - 2 187 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY712603.1 - 1 - 2 53 Chr. 11 - 117611380 - 117612720 [+] on Build GRCm38 70
          GenBankmRNA BB632597.1 - 1 - 2 90 Chr. 11 - 117611345 - 117613535 [+] on Build GRCm38 70
          GenBankmRNA BB645072.1 - 1 - 2 187 Chr. 11 - 117611247 - 117612736 [+] on Build GRCm38 70
          GenBankmRNA BB632596.1 - 1 - 2 90 Chr. 11 - 117611345 - 117613541 [+] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 11732 Gm11732 OTTMUSG00000003800 -172.4kb Sense Chr. 11 - 117426163 - 117438866 on NCBI Build GRCm38
          Downstream trinucleotide repeat containing 6C Tnrc6c 9930033H14Rik,
          mKIAA1582
          40.9kb Sense Chr. 11 - 117654289 - 117763439 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001964.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062314 NR_015489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062314 AK019332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062314 AK046435.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179518 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062314 NR_015489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062314 AK019332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062314 AK046435.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179518 - -
          Whole Mouse Genome G4122A A_52_P13936 AK038849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P13936 AK038850.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119209 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09450190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09638811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09731573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09366735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09215970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09164043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09229324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09224077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09609669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09370924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09366729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09742863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09727299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09097610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09224080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28166142 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444801_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382932 - -
          Mouse Genome 430 2.0 Array 1444801_at - -
          Mouse Genome 430 2.0 Array 1444801_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100018693.mm.1 TC1100001964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018693.mm.1 NR_015489.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018692.mm.1 TC1100001964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018692.mm.1 NR_015489.2 Refseq
          Mouse Expression Set 430 1444801_at - -
          Mouse Expression Set 430 1444801_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444801_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444801_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 314724 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106590735 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0