Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206551_gH
          See other GM10094 GE Assays ›
          Gene Symbol:
          Gm10094 Sap18
          Gene Name:
          predicted gene 10094
          Sin3-associated polypeptide 18
          Gene Aliases:
          ENSMUSG00000061104
          C530046K05Rik, D11Ertd539e, EMegR4, Sinbp1
          Chromosome Location:
          Chr.8: 95825356 - 95826156 on Build GRCm38
          Chromosome Location:
          Chr.14: 57798189 - 57804980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm04206551_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10094
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142441.1 NP_001135913.1 1 368 104
          Gene Symbol: Sap18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009119.3 NP_033145.2 3 - 4 397 104
          GenBank mRNA AK005189.1 - 3 - 4 315 104
          AK006647.1 - 3 - 4 336 104
          AK085992.1 - 3 - 4 336 104
          AK133619.1 - 3 - 4 331 104
          AK144904.1 - 3 - 4 370 104
          AK146928.1 - 3 - 4 370 104
          BC006625.1 - 3 - 4 338 3968600 104
          BC099480.1 - 3 - 4 331 30934194 104
          CF730347.1 - 3 - 4 397 104
          Z97062.1 - 3 - 4 332 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000825.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm10094
          Click here to find siRNAs for Gene Sap18

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of alternative mRNA splicing, via spliceosome
          chromatin organization
          regulation of transcription, DNA-templated
          positive regulation of apoptotic process
          negative regulation of mRNA splicing, via spliceosome
          molecular_function
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956195 CO797678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956195 NM_001142441.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956195 Z97062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956195 BC006625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956195 BC099480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956195 CO797678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956195 NM_001142441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956195 Z97062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956195 BC006625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956195 BC099480.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574414 - -
          Affymetrix Mouse Gene 1.1 ST Array 10574412 - -
          Affymetrix Mouse Clariom S Array PSR1400006770.mm.1 TC1400000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006770.mm.1 NM_001142441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006770.mm.1 NM_009119.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006770.mm.1 NM_009119.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006769.mm.1 TC1400000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006769.mm.1 NM_001142441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006769.mm.1 NM_009119.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006769.mm.1 NM_009119.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006774.mm.1 TC1400000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006774.mm.1 NM_009119.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006774.mm.1 NM_009119.3 Refseq
          Mouse Expression Set 430 1419444_PM_at - -
          Mouse Expression Set 430 1419444_at - -
          Mouse Expression Set 430 1419445_PM_s_at - -
          Mouse Expression Set 430 1434646_s_at - -
          Mouse Expression Set 430 1419445_s_at - -
          Mouse Expression Set 430 1434646_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1419445_s_at - -
          Mouse Genome 430A 2.0 Array 1419444_PM_at - -
          Mouse Genome 430A 2.0 Array 1419445_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1434646_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1434646_s_at - -
          Mouse Genome 430A 2.0 Array 1419444_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-b98c95889-pjv49:80/100.66.134.79:80.
          git-commit: ef20cda39f797f656ba0530dba66184d0857c864
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0