Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04204484_g1
          See other PKIG GE Assays ›
          Gene Symbol:
          Pkig
          Gene Name:
          protein kinase inhibitor, gamma
          Gene Aliases:
          PKIgamma
          Chromosome Location:
          Chr.2: 163658386 - 163726158 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04204484_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pkig
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039390.2 NP_001034479.1 3 - 4 262 108
          NM_001039391.2 NP_001034480.1 2 - 3 176 108
          NM_001164053.1 NP_001157525.1 2 - 3 271 108
          NM_001277096.1 NP_001264025.1 2 - 3 176 108
          NM_011106.2 NP_035236.1 2 - 3 189 108
          GenBank mRNA AK135505.1 - 2 - 3 177 108
          AK164716.1 - 2 - 3 164 108
          AK168614.1 - 2 - 3 114 108
          BC026550.1 - 2 - 3 174 5006826 108
          BY240874.1 - 2 - 3 241 108
          BY266193.1 - 3 - 4 358 108
          BY283174.1 - 3 - 4 197 108
          BY343638.1 - 2 - 3 153 108
          CJ051558.1 - 2 - 3 177 108
          U97170.1 - 2 - 3 193 108
          W75470.1 - 2 - 3 203 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002494.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK164716.1, AK168614.1, BC026550.1, NM_001039390.2, NM_001039391.2, NM_001164053.1, NM_001277096.1, NM_011106.2, U97170.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pkig

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          negative regulation of protein kinase activity
          signal transduction
          negative regulation of protein import into nucleus
          negative regulation of protein serine/threonine kinase activity
          negative regulation of cAMP-dependent protein kinase activity
          protein kinase inhibitor activity
          cAMP-dependent protein kinase inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 TC0200002494.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001039390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001039390.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001039391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001039391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001164053.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001164053.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001164055.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001164055.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_001277096.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_011106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020360.mm.1 NM_011106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 TC0200002494.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001039390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001039390.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001039391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001039391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001164053.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001164053.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001164055.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001164055.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_001277096.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_011106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020364.mm.1 NM_011106.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header