Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04178668_mH
          See other RHOX3A GE Assays ›
          Gene Symbol:
          Rhox3a Rhox3c Rhox3e Rhox3f Rhox3g Rhox3h
          Gene Name:
          reproductive homeobox 3A
          reproductive homeobox 3C
          reproductive homeobox 3E
          reproductive homeobox 3F
          reproductive homeobox 3G
          reproductive homeobox 3H
          Gene Aliases:
          AY147207, Gm1138, Prx, Rhox3, Rhox3.1
          EG665203, Rhox3.3, Rhox3b
          ENSMUSG00000071772, OTTMUSG00000017148, Rhox3.5, Rhox3d
          OTTMUSG00000017155, Rhox3.6, Rhox3e
          OTTMUSG00000017157, Rhox3.7, Rhox3f, Rhox3g-ps
          EG434758, Rhox3.8, Rhox3g
          Chromosome Location:
          Chr.X: 37249919 - 37258978 on Build GRCm38
          Chromosome Location:
          Chr.X: 37469868 - 37473961 on Build GRCm38
          Chromosome Location:
          Chr.X: 37546975 - 37550897 on Build GRCm38
          Chromosome Location:
          Chr.X: 37581352 - 37585496 on Build GRCm38
          Chromosome Location:
          Chr.X: 37623431 - 37628139 on Build GRCm38
          Chromosome Location:
          Chr.X: 37657688 - 37668764 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          Assay ID Mm04178668_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhox3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_194063.3 NP_918952.2 4 - 5 769 86
          GenBank mRNA DQ058640.1 - 4 - 5 769 86
          Gene Symbol: Rhox3c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001102457.4 NP_001095927.3 2 - 3 549 86
          Gene Symbol: Rhox3e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001184969.1 NP_001171898.1 549 86
          Gene Symbol: Rhox3f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040089.3 NP_001035178.3 2 - 3 549 86
          GenBank mRNA AY147207.1 - 442 86
          BC145759.1 - 419 86
          Gene Symbol: Rhox3g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145406.3 NP_001138878.1 419 86
          GenBank mRNA BC147475.1 - 419 9056117 86
          Gene Symbol: Rhox3h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114157.1 NP_001107629.1 4 - 5 769 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000339.mm.1, TC0X00000351.mm.1, TC0X00000353.mm.1, TC0X00003392.mm.1, TC0X00003423.mm.1, TC0X00003424.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhox3a
          Click here to find siRNAs for Gene Rhox3c
          Click here to find siRNAs for Gene Rhox3e
          Click here to find siRNAs for Gene Rhox3f
          Click here to find siRNAs for Gene Rhox3g
          Click here to find siRNAs for Gene Rhox3h

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00002410.mm.1 TC0X00000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002410.mm.1 NM_001184969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002410.mm.1 NM_001184969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002368.mm.1 TC0X00003392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002368.mm.1 NM_001102457.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002428.mm.1 TC0X00000353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002428.mm.1 NM_001040089.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002428.mm.1 NM_001040089.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013153.mm.1 TC0X00003424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00013153.mm.1 NM_001114157.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013153.mm.1 NM_001114157.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002255.mm.1 TC0X00000339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002255.mm.1 NM_194063.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002255.mm.1 NM_194063.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002369.mm.1 TC0X00003392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002369.mm.1 NM_001102457.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002411.mm.1 TC0X00000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002411.mm.1 NM_001184969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002411.mm.1 NM_001184969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002427.mm.1 TC0X00000353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002427.mm.1 NM_001040089.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002427.mm.1 NM_001040089.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013154.mm.1 TC0X00003424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00013154.mm.1 NM_001114157.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013154.mm.1 NM_001114157.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002254.mm.1 TC0X00000339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002254.mm.1 NM_194063.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002254.mm.1 NM_194063.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002408.mm.1 TC0X00000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002408.mm.1 NM_001184969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002408.mm.1 NM_001184969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002409.mm.1 TC0X00000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002409.mm.1 NM_001184969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002409.mm.1 NM_001184969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002412.mm.1 TC0X00000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002412.mm.1 NM_001184969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002412.mm.1 NM_001184969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013145.mm.1 TC0X00003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00013145.mm.1 NM_001145406.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-69f987f899-9tng4:80/100.66.130.250:80.
          git-commit: eb1b8c2b8eec225754209f035b16fef014668c1b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0