Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03979096_m1
          See other GM15958 GE Assays ›
          Gene Symbol:
          Gm15958
          Gene Name:
          predicted gene 15958
          Gene Aliases:
          OTTMUSG00000028223
          Chromosome Location:
          Chr.18: 66312259 - 66319608 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03979096_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15958
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK138348.1 - 702 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172906 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156761 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149054 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09034049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156761 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13771253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14045354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13685698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13771250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13960278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13617634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31786300 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1441784_PM_at - -
          Mouse Genome 430 2.0 Array 1441784_at - -
          Mouse Expression Set 430 1441784_at - -
          Mouse Expression Set 430 1441784_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0