Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03975992_g1
          See other GM19784 GE Assays ›
          Gene Symbol:
          Gm19784
          Gene Name:
          predicted gene, 19784
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 66924762 - 66949961 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03975992_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19784
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040461.1 4 - 5 533 70
          GenBank mRNA AK136089.1 - 1 - 2 181 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY720385.1 - 4 - 5 534 Chr. 18 - 66925427 - 66949961 [-] on Build GRCm38 70
          GenBankmRNA CJ052949.1 - 1 - 2 181 Chr. 18 - 66925143 - 66926373 [-] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream melanocortin 4 receptor Mc4r Mc4-r -64.3kb Sense Chr. 18 - 66857705 - 66860487 on NCBI Build GRCm38
          Downstream guanine nucleotide binding protein,
          alpha stimulating,
          olfactory type
          Gnal 2610011C15Rik,
          9630020G10Rik,
          Galphaolf,
          Gna10,
          Golf
          161.9kb Anti-sense Chr. 18 - 67088298 - 67226792 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001486.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_52_P296452 AK136089.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13602045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13649747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14050991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13602051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14086698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13703415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13677504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14074538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31789074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13989180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800010956.mm.1 TC1800001486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010956.mm.1 NR_040461.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010956.mm.1 NR_040461.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010959.mm.1 TC1800001486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010959.mm.1 NR_040461.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010959.mm.1 NR_040461.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010960.mm.1 TC1800001486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010960.mm.1 NR_040461.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010960.mm.1 NR_040461.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010958.mm.1 TC1800001486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010958.mm.1 NR_040461.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010958.mm.1 NR_040461.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010955.mm.1 TC1800001486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010955.mm.1 NR_040461.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010955.mm.1 NR_040461.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0