Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03958578_s1
          See other REDRUM GE Assays ›
          Gene Symbol:
          Redrum
          Gene Name:
          Redrum, erythroid developmental long intergenic non-protein coding transcript
          Gene Aliases:
          9430076G02Rik
          Chromosome Location:
          Chr.18: 54422295 - 54453294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03958578_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Redrum
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040338.1 3 2160 105
          GenBank mRNA AK020489.1 - 1 943 105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY427571.1 - 1 329 Chr. 18 - 54452898 - 54453289 [+] on Build GRCm38 105
          GenBankmRNA BY589440.1 - 1 Chr. 18 - 54453000 - 54453294 [+] on Build GRCm38 105
          GenBankmRNA BY588759.1 - 1 381 Chr. 18 - 54452849 - 54453294 [+] on Build GRCm38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream casein kinase 1,
          gamma 3
          Csnk1g3 3300002K07Rik,
          C330049O21Rik
          -496.6kb Sense Chr. 18 - 53862082 - 53955684 on NCBI Build GRCm38
          Downstream RIKEN cDNA 9330117O12 gene 9330117O12Rik 9330117O12 158.5kb Sense Chr. 18 - 54611744 - 54639870 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000550.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2243566 AK036466.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385654 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155339 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2243566 AK036466.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09005779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03479570 - -
          Whole Mouse Genome G4122A A_52_P169696 AK036466.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14090170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13853103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13884027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13749680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13941161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13606093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14008713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13749699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13837687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13787670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13873743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13853104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13837698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13890910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13956775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13944008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14008705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13749709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14136631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13880752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13837697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13944018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13812667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13723599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13641514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14097041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13616297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14060204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13749712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31718833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430839_PM_at - -
          Mouse Genome 430 2.0 Array 1430839_at - -
          Mouse Genome 430 2.0 Array 1430839_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800003945.mm.1 TC1800000550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800003945.mm.1 NR_040338.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003945.mm.1 NR_040338.1 Refseq
          Mouse Expression Set 430 1430839_at - -
          Mouse Expression Set 430 1430839_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430839_at - -
          Affymetrix HT Mouse Genome 430B Array 1430839_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 802973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0