Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03955833_m1
          See other 4930412F09RIK GE Assays ›
          Gene Symbol:
          4930412F09Rik
          Gene Name:
          RIKEN cDNA 4930412F09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03955833_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930412F09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015119.1 - 2 - 3 135 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714665.1 - 2 - 3 130 Chr. 6 - 30896903 - 30943348 [+] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coatomer protein complex,
          subunit gamma 2
          Copg2 AW227625,
          gamma-2-COP
          -11.3kb Anti-sense Chr. 6 - 30809559 - 30896794 on NCBI Build GRCm38
          Downstream Kruppel-like factor 14 Klf14 5330411L03Rik,
          AL022736,
          BTEB5
          12.6kb Anti-sense Chr. 6 - 30956021 - 30958990 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2408175 AK015119.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125406 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2408175 AK015119.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05850941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05850913 - -
          Whole Mouse Genome G4122A A_51_P518987 AK015119.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123771 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05850941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05640282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05849781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24334648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431635_PM_at - -
          Mouse Genome 430 2.0 Array 1431635_PM_at - -
          Mouse Genome 430 2.0 Array 1431635_at - -
          Mouse Expression Set 430 1431635_PM_at - -
          Mouse Expression Set 430 1431635_at - -
          Affymetrix HT Mouse Genome 430B Array 1431635_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431635_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0