Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952983_s1
          See other PANTR2 GE Assays ›
          Gene Symbol:
          Pantr2
          Gene Name:
          POU domain, class 3, transcription factor 3 adjacent noncoding transcript 2
          Gene Aliases:
          2610207O16Rik, Pou3f3os2, linc-Brn1b
          Chromosome Location:
          Chr.1: 42707061 - 42713454 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952983_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pantr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110495.1 4 319 96
          GenBank mRNA AK011788.1 - 1 848 96
          AK011913.1 - 1 848 96
          AW488209.1 - 1 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 2900092D14 gene 2900092D14Rik -3.9kb Anti-sense Chr. 1 - 42700336 - 42703176 on NCBI Build GRCm38
          Downstream mitochondrial ribosomal protein S9 Mrps9 2310002A08Rik,
          AW322516,
          Mrp-S9,
          S9mt,
          mg637
          143.2kb Anti-sense Chr. 1 - 42851233 - 42905683 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002258.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s)

          cerebellar cortex structural organization
          multicellular organism growth
          homeostasis of number of cells within a tissue

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01099183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00267526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00364381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01025625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01099194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01099179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01099181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20184788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00020197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01025628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00853261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00735299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01025626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100018147.mm.1 TC0100002258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018147.mm.1 NR_110495.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018148.mm.1 TC0100002258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018148.mm.1 NR_110495.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 543033 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104010347 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0