Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03950192_m1
          See other LOC100036513 GE Assays ›
          Gene Symbol:
          LOC100036513
          Gene Name:
          uncharacterized LOC100036513
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03950192_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100036513
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK151815.1 - 2 - 3 265 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY744567.1 - 2 - 3 265 Chr. 3 - 7612705 - 7689097 [+] on Build GRCm38 75
          GenBankmRNA BY164440.1 - 2 - 3 265 Chr. 3 - 7612705 - 7688778 [+] on Build GRCm38 75
          GenBankmRNA BY164177.1 - 2 - 3 265 Chr. 3 - 7612705 - 7688766 [+] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream interleukin 7 Il7 A630026I06Rik,
          Il-7,
          hlb368
          -50.2kb Anti-sense Chr. 3 - 7572028 - 7613427 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700010I02 gene 1700010I02Rik 235.3kb Anti-sense Chr. 3 - 7925303 - 7954887 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395742 AK037084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735410 AK080902.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73727 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02715396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03036025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03145987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02603932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02656563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21929931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0