Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949737_m1
          See other C78283 GE Assays ›
          Gene Symbol:
          C78283
          Gene Name:
          expressed sequence C78283
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949737_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C78283
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK144862.1 - 1 - 2 247 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV468321.1 - 1 - 2 227 Chr. 5 - 73946402 - 73948039 [+] on Build GRCm38 63
          GenBankmRNA BY734523.1 - 1 - 2 245 Chr. 5 - 73946383 - 73948259 [+] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700019F05 gene 1700019F05Rik -26.0kb Sense Chr. 5 - 73912889 - 73920417 on NCBI Build GRCm38
          Downstream ubiquitin specific peptidase 46 Usp46 1190009E20Rik,
          2410018I08Rik,
          AI451644
          50.4kb Anti-sense Chr. 5 - 73998726 - 74068411 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2208866 C78283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2208866 AK144862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2208866 BG066164.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112287 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04592519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04687454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04729408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04843041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04338994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04338990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04687452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04523583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23800452 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-mvvkb:80/100.66.135.185:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0