Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949639_s1
          See other GID8 GE Assays ›
          Gene Symbol:
          Gid8
          Gene Name:
          GID complex subunit 8 homolog (S. cerevisiae)
          Gene Aliases:
          2310003C23Rik, 4833420G11Rik, AI451474, Twa1
          Chromosome Location:
          Chr.2: 180710226 - 180721599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949639_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gid8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289651.1 NP_001276580.1 5 4367 93
          NM_001289652.1 NP_001276581.1 5 4333 93
          NM_029607.2 NP_083883.1 4 4853 93
          GenBank mRNA AK143107.1 - 1 2153 93
          BQ175265.1 - 1 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI661911.1 - 440 Chr. - - [-] on Build 93
          GenBankmRNA AI846776.1 - 1 Chr. 2 - 180721228 - 180721599 [-] on Build GRCm38 93
          GenBankmRNA BI499319.1 - 2 Chr. 2 - 180721154 - 180734435 [-] on Build GRCm38 93
          GenBankmRNA BB091185.1 - 1 224 Chr. 2 - 180721297 - 180721595 [+] on Build GRCm38 93
          GenBankmRNA BB025945.1 - 1 229 Chr. 2 - 180721304 - 180721597 [+] on Build GRCm38 93
          GenBankmRNA BI319845.1 - 1 Chr. 2 - 180721244 - 180721599 [-] on Build GRCm38 93
          GenBankmRNA CF577539.1 - 2 Chr. 2 - 180721159 - 180734435 [-] on Build GRCm38 93
          GenBankmRNA CD773907.1 - 1 Chr. 2 - 180720949 - 180721594 [-] on Build GRCm38 93
          GenBankmRNA BE634773.1 - 1 Chr. 2 - 180721155 - 180721599 [-] on Build GRCm38 93
          GenBankmRNA BQ747196.1 - 200 Chr. - - [-] on Build 93
          GenBankmRNA DV654352.1 - 1 448 Chr. 2 - 180721077 - 180721567 [+] on Build GRCm38 93
          GenBankmRNA BU842766.1 - 282 Chr. - - [-] on Build 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream death inducer-obliterator 1 Dido1 6720461J16Rik,
          D130048F08Rik,
          DATF-1,
          DIO-1,
          Datf1,
          dido
          -9.3kb Anti-sense Chr. 2 - 180657963 - 180710035 on NCBI Build GRCm38
          Downstream solute carrier family 17,
          member 9
          Slc17a9 1700019H03Rik,
          BC019537,
          Vnut
          3.6kb Sense Chr. 2 - 180725213 - 180742280 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gid8

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2398534 AA277533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2398534 AK143107.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262521 - -
          Agilent Mouse cDNA Microarray (G4104A) 764342 - -
          Agilent Mouse cDNA Microarray (G4104A) 1209243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1455254_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455255_PM_at - -
          Mouse Genome 430 2.0 Array 1455254_at - -
          Mouse Genome 430 2.0 Array 1455254_PM_at - -
          Mouse Genome 430 2.0 Array 1455255_at - -
          Mouse Genome 430 2.0 Array 1455255_PM_at - -
          Mouse Expression Set 430 1455255_PM_at - -
          Mouse Expression Set 430 1455254_PM_at - -
          Mouse Expression Set 430 1455254_at - -
          Mouse Expression Set 430 1455255_at - -
          Murine Genome U74v2 Set 128578_r_at - -
          Murine Genome U74v2 Set 128578_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1455255_at - -
          Mouse Genome 430A 2.0 Array 1455254_at - -
          Mouse Genome 430A 2.0 Array 1455255_PM_at - -
          Mouse Genome 430A 2.0 Array 1455254_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455254_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455255_at - -
          Affymetrix HT Mouse Genome 430A Array 1455255_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455254_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 686591 - -
          ABI Mouse Genome Survey Microarray 922491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6b8df48494-rmf8w:80/100.66.134.75:80.
          git-commit: 2c12d7358bbebd854ca1589f3ab2f515852a3cea
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0