Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949513_m1
          See other GM1627 GE Assays ›
          Gene Symbol:
          Gm1627
          Gene Name:
          predicted gene 1627
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949513_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm1627
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK142343.1 - 1 - 2 151 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY730468.1 - 1 - 2 147 Chr. 1 - 133962165 - 133970003 [+] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream proline arginine-rich end leucine-rich repeat Prelp 7330409J17Rik,
          SLRR2A
          -32.2kb Anti-sense Chr. 1 - 133910304 - 133929976 on NCBI Build GRCm38
          Downstream fibromodulin Fmod AI131919,
          AU041740,
          SLRR2E
          65.6kb Sense Chr. 1 - 134037515 - 134048277 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00320902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00136124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01009693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00476237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00157651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00392000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00392005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00391996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00157652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00595313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00210509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00787241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00157656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00476234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00320903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00141844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20630516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00392006 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0