Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949053_g1
          See other AU015791 GE Assays ›
          Gene Symbol:
          AU015791
          Gene Name:
          expressed sequence AU015791
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 105513433 - 105515249 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949053_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AU015791
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK139027.1 - 1 - 2 980 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV381755.2 - 2 - 3 193 Chr. 12 - 105513734 - 105515246 [+] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA D430019H16 gene D430019H16Rik AI852661 -20.4kb Sense Chr. 12 - 105453856 - 105493095 on NCBI Build GRCm38
          Downstream bradykinin receptor,
          beta 2
          Bdkrb2 B(2),
          B2,
          B2R,
          BK-2,
          BK2,
          BK2R,
          BKR2,
          BRB2
          47.9kb Sense Chr. 12 - 105563172 - 105595244 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2331804 AK139027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2331804 AV381755.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10177862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10223700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10177867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09796715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10223693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10324402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10325240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09796713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10222094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09796717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10252073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10149201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10252074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10223691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1442012_PM_at - -
          Mouse Genome 430 2.0 Array 1442012_at - -
          Mouse Genome 430 2.0 Array 1442012_PM_at - -
          Mouse Expression Set 430 1442012_at - -
          Mouse Expression Set 430 1442012_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442012_at - -
          Affymetrix HT Mouse Genome 430B Array 1442012_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0