Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948595_m1
          See other GM7538 GE Assays ›
          Gene Symbol:
          Gm7538
          Gene Name:
          predicted gene 7538
          Gene Aliases:
          EG665194
          Chromosome Location:
          Chr.5: 119194313 - 119200018 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948595_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm7538
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046033.1 2 - 3 144 72
          GenBank mRNA AK135898.1 - 2 - 3 145 72
          AK162140.1 - 2 - 3 145 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY719825.1 - 2 - 3 145 Chr. 5 - 119195151 - 119200018 [-] on Build GRCm38 72
          GenBankmRNA CJ052772.1 - 2 - 3 145 Chr. 5 - 119195164 - 119200018 [-] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700081H04 gene 1700081H04Rik -79.8kb Anti-sense Chr. 5 - 119108236 - 119114543 on NCBI Build GRCm38
          Downstream predicted gene,
          31314
          Gm31314 273.5kb Sense Chr. 5 - 119472143 - 119527976 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003151.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998882 AK135898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998882 AK162140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24617 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998882 AK135898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998882 AK162140.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01201609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04619775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04459410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04459411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04118466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04944781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04118464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04133099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04390767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04619774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04619772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04619168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04234799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04385190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04439407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04118468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04622397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24031344 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500027950.mm.1 TC0500003151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027950.mm.1 NR_046033.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027950.mm.1 NR_046033.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0