Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948192_m1
          See other GM1653 GE Assays ›
          Gene Symbol:
          Gm1653
          Gene Name:
          predicted gene 1653
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 149274737 - 149279980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948192_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm1653
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040591.1 2 - 3 612 82
          GenBank mRNA AK133793.1 - 2 - 3 613 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB618725.1 - 2 - 3 613 Chr. 3 - 149274737 - 149278199 [+] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          36888
          Gm36888 -1.6mb Anti-sense Chr. 3 - 147620811 - 147635647 on NCBI Build GRCm38
          Downstream predicted gene 5149 Gm5149 EG381483 55.7kb Sense Chr. 3 - 149335731 - 149340637 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001512.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02468934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02739914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02585084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02382728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02585083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02834159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02382731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02589724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02913383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02947428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02468924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02286600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02612254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632507 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300012211.mm.1 TC0300001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012211.mm.1 NR_040591.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012211.mm.1 NR_040591.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0