Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03944367_m1
          See other 1700029M03RIK GE Assays ›
          Gene Symbol:
          1700029M03Rik
          Gene Name:
          RIKEN cDNA 1700029M03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 164960664 - 164961400 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03944367_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700029M03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK076808.1 - 126 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB615319.1 - 1 - 2 122 Chr. 1 - 164960924 - 164962825 [+] on Build GRCm38 87
          GenBankmRNA BB013992.2 - 1 - 2 126 Chr. 1 - 164960921 - 164962822 [+] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chemokine (C motif) ligand 1 Xcl1 AI661682,
          ATAC,
          LTN,
          Lptn,
          SCM-1,
          SCM-1a,
          Scyc1
          -25.4kb Anti-sense Chr. 1 - 164931648 - 164935510 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930568G15 gene 4930568G15Rik 103.8kb Anti-sense Chr. 1 - 165066617 - 165091930 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176653 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155760 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010582 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155760 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00700351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01057634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00832751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00700350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00650494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01012301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00133849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01037954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00133853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00650481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00133851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00703217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00133856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01012300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00703216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00036242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20792070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178492 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440793_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440794_PM_x_at - -
          Mouse Genome 430 2.0 Array 1440794_x_at - -
          Mouse Genome 430 2.0 Array 1440794_PM_x_at - -
          Mouse Genome 430 2.0 Array 1440793_PM_at - -
          Mouse Genome 430 2.0 Array 1440793_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69476_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69476_at - -
          Mouse Expression Set 430 1440793_PM_at - -
          Mouse Expression Set 430 1440794_PM_x_at - -
          Mouse Expression Set 430 1440793_at - -
          Mouse Expression Set 430 1440794_x_at - -
          Murine Genome U74v2 Set 170159_PM_at - -
          Murine Genome U74v2 Set 170159_at - -
          Affymetrix HT Mouse Genome 430B Array 1440794_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440794_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440793_at - -
          Affymetrix HT Mouse Genome 430B Array 1440793_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101580086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0