Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
TC0700003977.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_001242365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_199009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033070.mm.1 |
NM_199009.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
TC0700003977.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_001242365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_199009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033071.mm.1 |
NM_199009.3 |
Refseq |