Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03938566_s1
          See other 4930535I16RIK GE Assays ›
          Gene Symbol:
          4930535I16Rik
          Gene Name:
          RIKEN cDNA 4930535I16 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03938566_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930535I16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK050163.1 - 1 1047 102
          AK085801.1 - 1 1071 102
          BB667541.1 - 1 451 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF723499.1 - 1 158 Chr. 4 - 123915770 - 123916385 [-] on Build GRCm38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream c-myc binding protein Mycbp 5730488M09Rik,
          AW552132,
          AW742590,
          Amy-1
          -3.7kb Anti-sense Chr. 4 - 123905013 - 123912250 on NCBI Build GRCm38
          Downstream Ras-related GTP binding C Rragc AU041672,
          Gtr2,
          RAGC,
          TIB929,
          YGR163W
          159.0 Anti-sense Chr. 4 - 123917433 - 123936997 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174240 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123346 - -
          Whole Mouse Genome G4122A A_52_P281909 AK015982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281909 AK050163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281909 AK085801.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130491 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123346 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1427534_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430608_PM_at - -
          Mouse Genome 430 2.0 Array 1427534_PM_at - -
          Mouse Genome 430 2.0 Array 1430608_PM_at - -
          Mouse Genome 430 2.0 Array 1430608_at - -
          Mouse Genome 430 2.0 Array 1427534_at - -
          Mouse Expression Set 430 1427534_PM_at - -
          Mouse Expression Set 430 1427534_at - -
          Mouse Expression Set 430 1430608_at - -
          Mouse Expression Set 430 1430608_PM_at - -
          Mouse Genome 430A 2.0 Array 1427534_at - -
          Mouse Genome 430A 2.0 Array 1427534_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427534_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427534_at - -
          Affymetrix HT Mouse Genome 430B Array 1430608_at - -
          Affymetrix HT Mouse Genome 430B Array 1430608_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100940131 - -
          Illumina mouse-6 v1.0 expression beadchip 103940176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255182 - -
          Illumina Sentrix mouse-6.v1 bead array 103940176 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103940176 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255182 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0